Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acriflavine Neutral, Spectrum™ Chemical
SDP

Catalog No. 18608436 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-608-436 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-608-436 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. AC180100GM
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 8048-52-0

Specifications

CAS Max % 1
pH Approx. 3.0
Packaging Amber Glass Bottle
Quantity 100 g
Grade Ungraded
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Acriflavine Neutral

CAUTION: For use by qualified personnel for research and development only. Read and understand the label and Material Safety Data Sheet (MSDS) prior to use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.