Learn More
Medchemexpress LLC HY-109020 10mg Medchemexpress, Acrizanib CAS:1229453-99-9 Purity:>98%
SDP
Catalog No. 501968649
Encompass_Preferred
Medchemexpress, HY-109020 10mg Acrizanib CAS:1229453-99-9 Acrizanib is a VEGFR-2 inhibitor, with an IC50 of 17.4 nM for BaF3-KDR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8649 Supplier Medchemexpress LLC Supplier No. HY10902010MG
Medchemexpress, HY-109020 10mg Acrizanib CAS:1229453-99-9 Acrizanib is a VEGFR-2 inhibitor, with an IC50 of 17.4 nM for BaF3-KDR. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.