Learn More
Cytiva Acro 50 Vent Devices with PTFE Membrane - 0.45 µm, hose barb (18/pkg)
Supplier:  Cytiva 4256
Encompass
50 mm Filters with Superior Performance for Demanding Applications•Reusable and autoclavable•PTFE membrane and polypropylene housing have broad chemical resistance•Large surface area provides greater throughput and high air flow rate•Available with a variety of inlet and outlet connectors This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3020