Learn More
Andwin Scientific ACRO 50 PTFE 1 UM 18/PK
SDP
Supplier:  Andwin Scientific 4258
Encompass_Preferred
The 1 m Acro vent filter can be used for several laboratory procedures, including venting bioreactors, fermentation tanks, and carboys and filtering agressive solvents and sterile gas purging of culture vessels. This product is autoclavable at a temperature of 121C (250F) for half an hour or 30 minutes. It is also reusable and has a huge surface area that can give a greater throughput. It is sold in a pack of 18.
Catalog No. 50-121-4514