Learn More
Cytiva Acrodisc Syringe Filters with Nylon Membrane - 0.2 µm, 13mm, male slip luer outlet (100/pkg 300/cs)
Supplier:  Cytiva 4427
Encompass
Acrodisc Syringe Filters with Nylon Membrane - 0.2 µm, 13mm, male slip luer outlet.Syringe filters with nylon membrane are ideal for the filtration of aqueous and solvent-based sample prep for HPLC (0.45 μm) and UHPLC (0.2 μm) applications. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending the life of chromatography columns and reducing system downtime.Nylon syringe filters show excellent chemical compatibility with esters, bases and alcohols. Nylon syringe filters are not recommended for use with strongly acidic fluids.13 mm Acrodisc syringe filters are designed for the filtration of sample volumes up to 10 mL. The syringe filters are HPLC certified for low levels of UV-absorbing extractables, ensuring accurate and reliable results. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3044