Learn More
Cytiva (Formerly Pall Lab) Acrodisc™ One Syringe Filters with wwPTFE membrane - 0.2 m (1000/pkg)
Supplier:  Cytiva AP4912
Encompass
wwPTFE membrane syringe filters are ideal for the filtration of a wide variety of common solvents including aqueous and organic solvents used in HPLC (0.45 m) and UHPLC (0.2 m) sample prep. Filtration protects sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.Large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL. Acrodisc PSF Syringe Filters are designed to be fully compatible with automated robotic handling systems offering reliable and worry-free operation.The membrane and housing are extremely low in levels of potential extractables. The syringe filters are HPLC certified for low levels of UV-absorbing extractables, ensuring accurate results. The syringe filters are low in protein and API binding which make them ideal for drug studies. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3430