Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filter with Versapor Membrane - 1.2 m, 25 mm modified acrylic housing (75/pkg 300/cs)
Supplier:  Cytiva 4488
Encompass
Non sterile syringe filters with Versapor membrane can be used for the filtration, clarification and prefiltration of general aqueous solutions. Versapor is an acrylic copolymer on a non-woven support membrane. The hydrophilic membrane offers excellent throughput with high liquid flow rates making these syringe filters ideal for clarification, prefiltration or the filtration of high particulate laden samples.The 25 mm syringe filter is ideal for processing sample volumes up to 100 mL. For research use only. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3054