Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Aqueous Filtration - 350UL, 0.2 m Supor membrane (10/pkg)
Supplier:  Cytiva 8019
Encompass
Fast Processing with Efficient Removal of Particulates New well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volumeNew outlet tip geometry facilitates direct flow of samples into receiver plate without concerns of cross-contaminationVaried membrane and pore size selection offers efficient particulate removalManufactured in accordance with SBS guidelines, allowing plates to be run in manual, semi-automated and automated processesSmooth top surface and textured window allow for easy labeling on the plates, as well as provide easy usage orientation through the A1 corner notch This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3147