Learn More
ACROBiosystems ACROBiosystems Claudin-9 Full Length Protein, VLP, HEK293, 95%, 20 μg
Catalog No. NC3948032
Encompass_Preferred
Human Claudin-9 Full Length Protein is presented as envelope virus-like particles (eVLPs), expressed in human HEK293 cells. These VLPs incorporate target membrane proteins into the lipid bilayer during budding, preserving authentic protein conformation, post-translational modifications, and orientation in a detergent-free environment. This product is ideal for immunization, binding assays such as ELISA, and cell-based assays including FACS.
  • Expressed as Met 1 - Val 217 sequence (Accession # O95484-1)
  • Highly immunogenic with repetitive multivalent display
  • Purity of 95% with endotoxin levels less than 1.0 EU/μg
  • Formulated in PBS, Arginine, pH7.4 with trehalose as protectant
  • Mean peak VLP radius of 60-85 nm determined by DLS
  • Supplied as 0.2 μm filtered liquid
  • Shipped on dry ice with storage at -70°C
Catalog No. NC3948032 Supplier ACROBiosystems Supplier No. CL9H52P520UG
Human Claudin-9 Full Length Protein is presented as envelope virus-like particles (eVLPs), expressed in human HEK293 cells. These VLPs incorporate target membrane proteins into the lipid bilayer during budding, preserving authentic protein conformation, post-translational modifications, and orientation in a detergent-free environment. This product is ideal for immunization, binding assays such as ELISA, and cell-based assays including FACS.
  • Expressed as Met 1 - Val 217 sequence (Accession # O95484-1)
  • Highly immunogenic with repetitive multivalent display
  • Purity of 95% with endotoxin levels less than 1.0 EU/μg
  • Formulated in PBS, Arginine, pH7.4 with trehalose as protectant
  • Mean peak VLP radius of 60-85 nm determined by DLS
  • Supplied as 0.2 μm filtered liquid
  • Shipped on dry ice with storage at -70°C