Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems ACROBiosystems Human IFN-γ ELISA Kit, PRO - 96 Tests (CEA-C006-96tests)

Catalog No. NC3802742
Encompass_Preferred

Ready-to-use sandwich ELISA kit for quantitative detection of human interferon-gamma (IFN-γ) in serum, plasma, and cell culture supernatants. The kit includes calibrated standards, high-quality antibody pairs, and complete reagents to perform 96 determinations with a breakable 96-well plate format.

  • Ready-to-use sandwich ELISA for quantitative detection of human IFN-γ.
  • Includes calibrated standards traceable to NIBSC/WHO and complete reagent set.
  • Sensitivity (limit of detection) 4 pg/mL with dynamic range 7.81 pg/mL-1000 pg/mL.
  • Sample types: serum, plasma, and cell culture supernatant.
  • Sample volume 50 μL per well.
  • Assay time approximately 2 hr.
  • 96-well plate format breakable into 12 x 8 well strips.
  • Unopened storage at 2-8 °C; opened kit components shelf life per kit labeling.
  • For research use only.

Catalog No. NC3802742 Supplier ACROBiosystems Supplier No. CEAC00696TESTS
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ready-to-use sandwich ELISA kit for quantitative detection of human interferon-gamma (IFN-γ) in serum, plasma, and cell culture supernatants. The kit includes calibrated standards, high-quality antibody pairs, and complete reagents to perform 96 determinations with a breakable 96-well plate format.

  • Ready-to-use sandwich ELISA for quantitative detection of human IFN-γ.
  • Includes calibrated standards traceable to NIBSC/WHO and complete reagent set.
  • Sensitivity (limit of detection) 4 pg/mL with dynamic range 7.81 pg/mL-1000 pg/mL.
  • Sample types: serum, plasma, and cell culture supernatant.
  • Sample volume 50 μL per well.
  • Assay time approximately 2 hr.
  • 96-well plate format breakable into 12 x 8 well strips.
  • Unopened storage at 2-8 °C; opened kit components shelf life per kit labeling.
  • For research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.