Learn More
ACROBiosystems ACROBiosystems R-PE Antibody Labeling Kit, 500 μg (ALK-A001-500ug)
Catalog No. NC3860666
Encompass_Preferred
R-PE Antibody Labeling Kit provides a rapid, simple workflow to covalently conjugate R-phycoerythrin (PE) to antibodies via modification of interchain disulfide bonds. The resulting PE-antibody conjugates are ready to use without further purification and are suitable for flow cytometry and other fluorescence-based assays.
  • Simple and quick conjugation of PE to antibody.
  • Conjugated antibody is ready to use without further purification.
  • Minimal antibody loss during labeling.
  • Includes modifier reagent, activated PE, and quencher reagent.
  • Produces PE conjugates with excitation at 488 nm/561 nm and emission at 575 nm.
  • Unopened kit should be stored at -20 °C; check outer packaging for expiration date.
Catalog No. NC3860666 Supplier ACROBiosystems Supplier No. ALKA001500UG
R-PE Antibody Labeling Kit provides a rapid, simple workflow to covalently conjugate R-phycoerythrin (PE) to antibodies via modification of interchain disulfide bonds. The resulting PE-antibody conjugates are ready to use without further purification and are suitable for flow cytometry and other fluorescence-based assays.
  • Simple and quick conjugation of PE to antibody.
  • Conjugated antibody is ready to use without further purification.
  • Minimal antibody loss during labeling.
  • Includes modifier reagent, activated PE, and quencher reagent.
  • Produces PE conjugates with excitation at 488 nm/561 nm and emission at 575 nm.
  • Unopened kit should be stored at -20 °C; check outer packaging for expiration date.