Learn More
Cytiva (Formerly Pall Lab) AcroCap™ Positive Pressure Devices - 0.2 m, gamm-irradiated (10/pkg)
Supplier:  Cytiva 4480
Encompass
AcroCap™ devices are presterilized, disposable filter units ideal for sterilization or clarification of up to 3 l of tissue culture media or other critical fluids.-Supor membrane provides high flow rates-Devices are self-venting via 0.02 m PTFE ventFilters are designed as positive pressure devices for use with a peristaltic pump or pressure vessel to eliminate the denaturation of proteins which may occur when using vacuum filtration devices.Ordering information: Individually packaged units come presterilized by gamma irradiation with ringstand clamp adapters and attached filling bell. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3049