Learn More
Cytiva (Formerly Pall Lab) Acrodisc PSF Syringe Filters with Nylon Membrane - 0.2 m (50/pkg 200/cs)
Supplier:  Cytiva AP4436
Encompass
Syringe filters with nylon membrane are ideal for the filtration of aqueous and solvent-based sample prep for HPLC (0.45 m) and UHPLC (0.2 m) applications. Filtration helps protect sensitive parts of analytical instrumentation from particulate build-up, extending chromatography column life and reducing system downtime.Nylon syringe filters show excellent chemical compatibility with esters, bases and alcohols. They are not recommended for use with strongly acidic fluids.Large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL Available with a built-in GxF prefilter for use with difficult-to-filter solutions.Acrodisc PSF Syringe Filters are designed to be fully compatible with automated robotic handling systems.HPLC certified for low levels of UV-absorbing extractables, ensuring accurate and reliable results. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1476522