Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filter - 10 m with Versapor membrane, polypropylene housing (50/pkg 200/cs)
Supplier:  Cytiva AP4001
Encompass
Syringe filters are used in analytical chemistry applications to remove particulate contamination prior to analysis. Filtering a sample prior to sample injection into analytical instrumentation will help protect sensitive parts of the instrument from particulate build-up and reduce system downtime. Acrodisc Syringe Filters with Versapor membranes are ideal for prefiltration and clarification applications. Making them useful for prefiltration of particulate laden samples, serum filtration, and dissolution testing. The devices are compatible for use with general aqueous and proteinaceous samples.Acrodisc PSF Syringe Filters feature a large effective filtration area (3.9 cm2) for increased sample throughput, ideal for sample volumes of 150 mL. Autopack tubes are designed to be fully compatible for use with automated robotic handling systems offering reliable and worry-free operation. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1050397