Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filter with Glass Fiber Filter - 1 m, 25 mm (1000/pkg)
Supplier:  Cytiva 4529
Encompass
Acrodisc Syringe Filters with Glass Fiber, 1 m (nominal).Syringe filters are used in analytical chemistry applications to remove particulate contamination prior to analysis. Filtering a sample prior to sample injection into analytical instrumentation will help protect sensitive parts of the instrument from particulate build-up and reduce system downtime. Acrodisc 25 mm syringe filters with glass fiber maximize throughput prefiltration of particulate-laden samples. They can be used alone or in series with another Acrodisc syringe filter.25 mm Standard Acrodisc Syringe Filters are ideal for sample volumes of 100 mL and feature an effective filtration area of 2.8 cm2. The 25 mm Standard Acrodisc Syringe Filters feature a polypropylene housing that is chemically resistant to harsh solvents and low in extractables. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0885943