Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Syringe Filter, PTFE, 13 mm, 0.45 μm, Nonpolar

Supplier:  Waters Corp WAT200502

Encompass_Preferred

The Waters Acrodisc syringe filter is designed for efficient sample preparation, featuring a nonpolar PTFE membrane.

  • Membrane material: PTFE (Polytetrafluoroethylene)
  • Pore size: 0.45 μm
  • Diameter: 13 mm
  • Polarity: Nonpolar
  • Quantity per pack: 100/pk
  • Compatible with THF and chloroform

Catalog No. 50-786-049


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.