Learn More
Waters Corp Waters Corporation Syringe Filter, PTFE, 13 mm, 0.45 μm, Non polar
Supplier:  Waters Corp WAT200821
Encompass_Preferred
Waters Corporation Syringe Filters, with hydrophobic PTFE membranes, enable efficient filtration of non-aqueous, organic, acidic, or basic samples. Featuring a 13 mm diameter and 0.45 μm pore size, these filters are HPLC accredited to ensure low extractables and high-quality analytical sample preparation.
  • Hydrophobic PTFE membranes for non-aqueous, organic, acidic, or basic samples.
  • HPLC accredited to ensure low extractables and high-quality filtration.
  • Polypropylene housing with PTFE membrane supports gas and organic solvent filtering.
  • Prevents “air bubble” lock in sample vials via 13 mm mini spike outlet.
  • Ensures correct pore size ratings for improved chromatography and instrument protection.
  • Provides consistent analytical results and low extractable components.
  • Processes sample volumes less than 10 mL.
Catalog No. 50-786-152