Learn More
Waters Corp Acrodisc Syringe Filter, PTFE, 25 mm, 0.45 μm
Supplier: Waters Corp WAT200500
Hydrophobic PTFE membranes are ideal for non-aqueous solvents, organic, highly acidic, or basic samples. Certified for HPLC, these filters ensure low extractables, high-quality analytical filtering, and are housed in polypropylene for gas and organic solvent filtration.
- Hydrophobic PTFE membranes for non-aqueous, organic, acidic, or basic solvents.
- HPLC certified for low extractables and high-quality analytical filtering.
- Validated for compatibility with typical HPLC solvents.
- Features low levels of UV-detectable extractables.
- Polypropylene housing ideal for gas and organic solvent filtration.
- Avoids “air bubble” lock with 13 mm tiny spike outlet.
- Superior separation technology for chromatography and instrument protection.