Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Acrodisc MiniSpike Syringe Filter, PTFE, 13 mm, 0.45 μm

Supplier:  Waters Corp WAT200825

Encompass_Preferred

This filter features hydrophobic, polytetrafluoroethene (PTFE) membranes suitable for use with non-aqueous solvents, organic-based, highly acidic, or basic samples and solvents.

  • Minispike syringe filter format
  • 13 mm diameter
  • Pore size of 0.45 μm
  • Hold-up volume less than 14 μL
  • Housing material is polypropylene
  • Recommended for sample volumes less than 10 mL
  • Non-polar separation mode

Catalog No. 50-786-156


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.