Learn More
Waters Corp Acrodisc, Minispike Syringe Filter, PTFE, 13 mm, 0.2 µm, Non polar, 100/pk
Supplier:  Waters Corp WAT200556
Encompass_Preferred
This PTFE membrane is a good choice for use with aggresive organic solvents
Catalog No. 50-786-091