Learn More
Waters Corp Waters Syringe Filter, PVDF, 13 mm, 0.2 μm, 100/pk
Supplier:  Waters Corp WAT200806
Encompass_Preferred
This Acrodisc Syringe Filter features hydrophilic polyvinylidene fluoride (PVDF) membranes, making it ideal for filtering both aqueous and organic solvents. It helps maintain optimal performance in chromatography systems by preventing particulate matter from settling in critical components.
  • Hydrophilic PVDF membranes are suitable for aqueous and organic solvents.
  • Filters buffers and materials, preventing particulate matter in columns and instruments.
  • Extends column lifespan and reduces system downtime.
  • Removes contaminants and degasses solvents in systems without on-line degassers.
  • Reduces particle wear on pistons, increases column life, and prevents pump downtime.
  • Used in biological sciences, pharmaceutical operations, R&D, quality control, and environmental applications.
  • Useful for pre-filtration, ion chromatography, HPLC, and UPLC.
Catalog No. 50-786-137