Learn More
Waters Corp Waters Syringe Filter, PVDF, 25 mm, 0.2 μm
Supplier:  Waters Corp WAT200808
Encompass_Preferred
The PVDF LC Syringe Filter maintains peak performance in laboratory analysis. It features a hydrophilic polyvinylidene fluoride (PVDF) membrane, making it suitable for both aqueous and organic liquids. These filters are versatile for use in biological sciences, pharmaceutical operations, research and development, quality control, and environmental applications.
  • Suitable for aqueous and organic liquids
  • Low protein binding for biological sample filtering
  • Extends column lifespan and reduces system downtime by preventing particulate contamination
  • Removes contaminants from solvents and degasses systems without on-line degassers
  • Used in pre-filtration, ion chromatography, high-pressure liquid chromatography, and ultra-high-pressure liquid chromatography
Catalog No. 50-786-139