Learn More
Waters Corp Waters Corporation Syringe Filter, PVDF, 25 mm, 0.45 μm
Supplier:  Waters Corp WAT200510
Encompass_Preferred
This syringe filter features a hydrophilic polyvinylidene fluoride (PVDF) membrane, suitable for aqueous and organic solvents. Designed for top performance, it supports high-pressure liquid chromatography, ultra-high-pressure liquid chromatography, ion chromatography, and pre-filtration. It prevents particulate matter accumulation, prolonging column lifespan and reducing system downtime by eliminating impurities and degassing solvents.
  • Hydrophilic PVDF membrane for aqueous and organic solvents
  • 25 mm diameter with 0.45 μm pore size
  • Ideal for biological, pharmaceutical, and R&D applications
  • Supports HPCL, UPLC, and ion chromatography
  • Prevents particulate accumulation, prolonging column life and reducing downtime
  • Low <100 μL hold-up volume, polypropylene housing
Catalog No. 50-786-057