Learn More
Waters Corp Waters Acrodisc Minispike Syringe Filter, PVDF, 0.2 μm, 13 mm
Supplier:  Waters Corp WAT200804
Encompass_Preferred
The Acrodisc Minispike Syringe Filter, featuring a hydrophilic polyvinylidene fluoride (PVDF) membrane, is ideal for filtering both aqueous and organic solvents. This lab equipment is crucial for various applications in biological sciences, pharmaceuticals, R&D, QC, and environmental studies, particularly for pre-filtration, ion chromatography, HPLC, and UHPLC.
  • Hydrophilic PVDF membrane for aqueous and organic solvents.
  • Supports biological sciences, pharmaceuticals, R&D, QC, and environmental applications.
  • Excellent for pre-filtration, ion chromatography, HPLC, and UHPLC.
  • Maintains peak performance, extends column lifespan, and reduces system downtime.
  • Filters buffers, removes contaminants, and degasses solvents for increased efficiency.
  • Prevents particulate settling, lessens piston wear, and increases column life.
Catalog No. 50-786-135