Learn More
Labsource Inc Acrodisc Mustang E Syringe Filter, Endotoxin Removal, Disposable Membrane, 10 Per Pack
SDP
Supplier:  Labsource Inc 28140521PK
Encompass_Preferred
Acrodisc Mustang E Syringe Filters are designed for rapid endotoxin removal from various solutions. With a 0.2 µm pore size and 25 mm diameter, these disposable filters ensure efficient method development and chemistry selection. They offer options for sterile or non-sterile use, making them versatile for laboratory needs. Beneficial in proof-of-concept studies, the filters can operate under varying temperatures and pressures, setting them apart as reliable chromatography units.
Catalog No. NC0509173