Learn More
Bellco Glass, Inc. AcroDisc Thread Filter, PKG/8
SDP
Supplier:  Bellco Glass, Inc. 196412922
Encompass_Preferred
The ptfe 50 mm Acrodisc hydrophobic filter with hose barbs can be used to sterilize non-aqueous solutions, vacuum line protection, venting and gas filtration. Hose barb end allows easy secure attachment of silicone tubing or other types of tubing. •ptfe acro fitler disc•package 8 in a 9x12 plastic bag•.02um size•with npt threads on each size of the filter•dimensions: 2 29/32" od x 2 1/4" long•non-sterile•autoclavable•do not use at pressures over 30 psig•color: natural•filtration area: 19.6cm2•filter diamter: 50mm•max operating temperature: 121°c•outside diam: 62mm•overall lenght: 71mm•outer material: polypropylene•filter material: hydrophobic ptfeAPPLICATIONS:•sterilize non-aquesous solutions•vacuum line protection•gas filtration•tank & bioreactor venting
Catalog No. 50-194-4770