Learn More
Chem Service Inc Acrolein, 107-02-8, MFCD00006998, 100 mg
SDP
Supplier:  Chem Service Inc N11030100MG
Encompass_Preferred
This item has a minimum order quantity of 2 per supplier requirements
Molecular Formula: C3 H4 O, Molecular Weight: 56.064, Alternate Name(s): 2-Propenal, INHIBITED WITH ~3% WATER & 1% CATECHOL, Density: 0.8389. Acrolein may be used as a reference standard in the determination of acrolein in human breast cancer cells using selected ion flow tube chemical ionization mass spectrometry.
Catalog No. NC1512384