Learn More
Medchemexpress LLC Acrolein 2,4-dinitrophenylhydrazone-3,5,6-d3 | 259824-62-9 | 99.95% | C9H5D3N4O4 | 1 MG
SDP
Catalog No. 5000354381
Encompass_Preferred
Acrolein 2,4-dinitrophenylhydrazone-3,5,6-d3 is the deuterium labeled Acrolein 2,4-dinitrophenylhydrazone-3,5,6, designed for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration can affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for drug molecule tracing
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-54381 Supplier Medchemexpress LLC Supplier No. HY141856S1MG
Acrolein 2,4-dinitrophenylhydrazone-3,5,6-d3 is the deuterium labeled Acrolein 2,4-dinitrophenylhydrazone-3,5,6, designed for research use only. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules as tracers for quantitation during drug development, and deuteration can affect pharmacokinetic and metabolic profiles.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Incorporates stable heavy isotopes for drug molecule tracing
  • Potential to affect pharmacokinetic and metabolic profiles of drugs