Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Acrolein 2,4-Dinitrophenylhydrazone 98.0+%, TCI America™
SDP

Catalog No. A12725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A12725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A12725G Supplier TCI America Supplier No. A12725G

missing translation for 'validMainframeMsg'

Specifications

CAS 888-54-0
Melting Point 165°C
Molecular Formula C9H8N4O4
MDL Number MFCD00191599
UN Number 1325
Quantity 5 g
Synonym acrolein 2,4-dinitrophenylhydrazone, acrolein-2,4-dinitrophenylhydrazone, 2-propenal, 2,4-dinitrophenyl hydrazone, acrolein-2,4-dinitrophenylhydrazone dnph, 2,4-dinitro-n-e-prop-2-enylideneamino aniline, 1e-1-azabuta-1,3-dienyl 2,4-dinitrophenyl amine, 1-allylidene-2-2,4-dinitrophenyl hydrazine, 2,4-dinitro-n-prop-2-enylideneamino aniline, 2-propenal, 2-2,4-dinitrophenyl hydrazone, acrolein, 2,4-dinitrophenyl hydrazone 8ci
InChI Key JJPZHGIYUVFTGG-BJMVGYQFSA-N
SMILES C=CC=NNC1=C(C=C(C=C1)[N+](=O)[O-])[N+](=O)[O-]
IUPAC Name 2,4-dinitro-N-[(E)-prop-2-enylideneamino]aniline
Molecular Weight (g/mol) 236.187
PubChem CID 9568683
Formula Weight 236.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Acrolein 2,4-Dinitrophenylhydrazone
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.