Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Acrolein dimethyl acetal

Catalog No. 1905435ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
19-054-35ML 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 19-054-35ML Supplier Sigma Aldrich Supplier No. 1905435ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Acrolein dimethyl acetal

Specifications

CAS 6044-68-4
Density 0.862 g/mL (at 25°C (literature))
Boiling Point 89°C to 90°C (lit.)
Molecular Formula C5H10O2
Refractive Index n20/D 1.395 (literature)
Linear Formula H2C=CHCH(OCH3)2
MDL Number MFCD00008632
Quantity 500 mg
Synonym 3,3-Dimethoxy-1-propene
Molecular Weight (g/mol) 102.13
Percent Purity 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.