Learn More
Thermo Scientific™ AcroMetrix SARS-CoV-2 Control (In-process, RUO)
Catalog No. 100954519 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Catalog No. Quantity
10-095-4519 Each
1 options
Catalog No. 10-095-4519 Supplier Thermo Scientific™ Supplier No. 954519
Add to Cart
Add to Cart
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix™ Coronavirus 2019 (COVID-19) RNA Control (RUO).
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix™ Coronavirus 2019 (COVID-19) RNA Control (RUO). This is a positive control kit containing two concentrations: a Low Positive and Ultra-Low Positive concentration to avoid false-negative results.
  • Full length genomic RNA from SARS-CoV-2
  • Kit contains one Low Positive control and one Ultra-low Positive control
  • Quantitated using Bio-Rad Droplet Digital™ PCR (ddPCR™)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C or lower
Description AcroMetrix Coronavirus 2019 (COVID-19) RNA Control (RUO)
Packaging One kit contains 2 vials per kit x 0.02mL per vial
Quantity Each
For Research Use Only.