Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ AcroMetrix Multi-Analyte SARS-CoV-2, Flu A/B, RSV A/B Control (RUO)

Catalog No. 954516 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
95-451-6 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 95-451-6 Supplier Thermo Scientific™ Supplier No. 954516
Add to Cart
Add to Cart
This item is not returnable. View return policy

Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19, Flu A/B and RSV A/B using the Thermo Scientific™ AcroMetrix™ Multi-Analyte SARS-CoV-2, Flu A/B, RSV A/B Control (RUO).

Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19, Flu A/B and RSV A/B using the AcroMetrix™ Multi-Analyte SARS-CoV-2 Control, Flu A/B, RSV A/B Control (RUO). This multiplex control can be used as a complete assay control starting from extraction to amplification and finally detection of COVID-19, Flu A/B and RSV A/B.

  • Inactivated SARS-CoV-2, Influenza A/B and Respiratory Syncytial Virus A/B in proprietary viral transport media
  • Full process control mimics a clinical sample
  • Multiplexed quantitative positive control
  • Kit contains 5 vials per kit x 1.5 mL per vial
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage -20°C
Description AcroMetrix Multi-Analyte SARS-CoV-2, Flu A/B, RSV A/B Control (RUO)
Packaging One kit contains 5 vials per kit x 1.5 mL per vial
Quantity 1 Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.