Learn More
Thermo Scientific™ AcroMetrix SARS-CoV-2 Control (Full-process, RUO)
Shop All Thermo Scientific ProductsDescription
Validate and monitor the performance of molecular diagnostic tests for detecting COVID-19 using the Thermo Scientific™ AcroMetrix SARS-CoV-2 Control (RUO). This control mimics a patient sample and can be used as a complete assay control starting from extraction to amplification and finally detection of COVID-19.
- Inactivated SARS-CoV-2 in proprietary viral transport media
- Full process control mimics a patient sample
- Qualitative positive control
- Kit contains 5 vials per kit x 1.5 mL per vial
Specifications
Specifications
| Content And Storage | -20°C |
| Description | AcroMetrix SARS-CoV-2 Control (RUO) |
| Packaging | One kit contains 5 vials per kit x 1.5 mL per vial |
| Quantity | 1 Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.