Learn More
Cytiva (Formerly Pall Lab) AcroPak™ 20 Filters with Supor Membrane - 0.8/0.2 m, gamma-irradiated, with filling bell (3/pkg)
Supplier:  Cytiva 12203
Encompass
Efficient Processing of Liquid Volumes Typically Up To 20 L-Fluorodyne II membrane is compatible with aqueous and many organic solvents-AcroPak 20 filters are designed to add convenience to small- and medium-volume filtrations-AcroPak 200 Fluorodyne II capsules provide fast processing of clean batches-Double layer sterilizing membrane enhances reliability-Upstream air vent prevents vapor lock-Manufactured without the use of adhesives to minimize extractables-Pre-sterilized by gamma irradiation This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1855082