Learn More
Cytiva (Formerly Pall Lab) AcroPak™ 200 Capsules with Supor EKV Membrane - 0.2 m, stepped hose barb inlet/stepped hose barb outlet with filling bell (3/pkg)
Supplier:  Cytiva 12094
Encompass
For Buffers, Tissue Culture Media, and Other Biological Fluids-Newer technology recommended for simplified scale-up-High filtration area and compact size are ideal for upscale trials-Minimal hold-up volume-Good wettability for reliable integrity test-Sealed membrane and housing uses fusion technology to eliminate potential extractables-Gamma sterilized, non-pyrogenic, and provided with a removable filling bell (except sanitary flange option)-Hydrophilic polyethersulfone membrane for low adsorption and wide chemical compatibility-Very high flow rates and consistently higher total solution throughputs due to superior porosity over other membranes This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3180