Learn More
Cytiva (Formerly Pall Lab) Acrodisc Syringe Filters with Posidyne membrane Optimized for Scale Up, 25 mm - 0.2 m , Sterile (50/pkg)
Supplier:  Cytiva 4908N
Encompass
Pall offer UpScale Acrodisc syringe filters with a range of different scaleable membrane chemistries:- Supor membrane has high flow rates and throughputs and is ideal for solutions where low protein binding is required. Not recommended with some ketones.- Fluorodyne II membrane offers high flow rates and is ideal for applications where PVDF membrane is specified. Not recommended with some ethers.- Ultipor membrane provides broad solvent and chemical compatibility, and low extractables.- Posidyne membrane enhances bioburden and pyrogen removal from aqueous solutions.The syringe filters are integrity testable (water bubble point) and have been bacterial retention tested to assure sterile filtrate.Applications: Drug development studies, determination of product, compatibility and recovery, preliminary filterability testing and small-volume liquid sterilization.This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3123