Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ 96 Filter Plates, 1 mL - 0.45 m, PTFE membrane (5/pkg)
Supplier:  Cytiva 5056N
Encompass
Optimized for a Variety of Applications Requiring Larger Per Well Volume ProcessingPlates are constructed of chemically resistant, biologically-inert polypropyleneRobotics-friendly design has single piece construction, designed in accordance with the ANSI/SBS X-2004 specificationsA serialized barcode label allows use in automated tracking systemsProprietary sealing technology individually seals each well, eliminating crosstalkAvailable in a variety of membrane configurationsExtended flow directors eliminate splattering and crosstalk of filtrate This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3128