Learn More
Cytiva AcroPrep™ Advance 96-well Filter Plates for Lysate Clearance - 350 µL, 3.0 µm Glass Fiber/1.2 µm Supor membrane (10/pkg)
Supplier:  Cytiva 8040
Encompass
Prefilter Provides Efficient Clarification of Highly Particulated Samples-Integrated prefilter yields consistent filtration of samples with high levels of gross particulate-New well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume-Innovative outlet tip geometry provides direct flow of samples into receiver plate without concerns of cross-contamination-Manufactured with biologically inert materials that allow clarification of most types of lysates without loss of target molecules- Ideal for the removal of bacterial and cellular debris prior to biomolecule purification This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC0873374