Learn More
Cytiva (Formerly Pall Lab) AcroPrep™ Advance 96-well Filter Plates for Lysate Clearance - 350 L, 3.0 m Glass Fiber/0.2 ?m Supor membrane (10/pkg)
Supplier: Cytiva 8075
Prefilter Provides Efficient Clarification of Highly Particulated Samples-Integrated prefilter yields consistent filtration of samples with high levels of gross particulate-New well geometry results in faster, more uniform filtration rates across the plate with reduced hold-up volume-Innovative outlet tip geometry provides direct flow of samples into receiver plate without concerns of cross-contamination-Manufactured with biologically inert materials that allow clarification of most types of lysates without loss of target molecules- Ideal for the removal of bacterial and cellular debris prior to biomolecule purification This product is not approved for use in medical, clinical, surgical or other patient applications.