Learn More
Enzo Life Sciences ACRP30 headless (mouse), (recombinant) (10µg)
SDP
Supplier:  Enzo Life Sciences ALX203003C010
Encompass_Preferred
Control compound for rmsMEGACD40L (ALX-522-120). Control compound for rmsMEGACD40L (ALX-522-120). Alternative name: Tail-Adp, Tail Adiponectin, Adipocyte complement related protein of 30kDa headless. Endotoxin: <0.1EU/µg protein (LAL-test). Purity: ≥90% (SDS-PAGE). MW: ~25kDa . Formulation: Lyophilized. Contains PBS. Source: Produced in HEK 293 cells. The collagen domain of mouse ACRP30 (aa 18-111) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG-tag. Long Term Storage: -20°C. Use/Stability: Stable for at least 6 months after receipt when stored as recommended. Handling: Avoid freeze/thaw cycles. After reconstitution, prepare aliquots and store at -20°C.
Catalog No. 50-200-8054