Learn More
Enzo Life Sciences ACRP30 headless (human) monoclonal antibody (Ne.Na) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804144C100
Encompass_Preferred
Clone: Ne.Na. Isotype: IgG1. Immunogen: Recombinant human ACRP30headless (tail adiponectin) (ALX-203-002). Source: Purified from concentrated hybridoma tissue culture supernatant. Species reactivity: Human. Applications: ELISA, IP, WB. Formulation: Liquid. In PBS containing 0.02% sodium azide.
Catalog No. 50-200-9794