Learn More
Thermo Scientific Thermostats (3)
Catalog No. NC1009146 Shop All Thermo Scientific Products
Encompass
4 millimeter anion exchange resin beads for water softening, nitrate and other anion removal from drinking and industrial water by exchanging chloride, nitrate and sulfate ions.
Catalog No. NC1009146 Supplier Thermo Scientific Supplier No. 085090
4 millimeter anion exchange resin beads for water softening, nitrate and other anion removal from drinking and industrial water by exchanging chloride, nitrate and sulfate ions.