Learn More
Aviva Systems Biology Corp ACRV1 Antibody - N-terminal region (ARP53590_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53590_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ACRV1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ACRV1. Alternate names: D11S4365, SP-10, SPACA2ApplicationWB. Species reactivity: Guinea Pig, Human, Rabbit
Catalog No. 50-014-61061