Learn More
Cayman Chemical Acrvastn 50mg
SDP
Catalog No. 502933229
Encompass_Preferred
A histamine H1 receptor antagonist (Ki = 10 nM in COS-7 cells expressing the human receptor); completely inhibits response to histamine in guinea pigs when administered at a dose of 1 mg/kg
Catalog No. 50-293-3229 Supplier Cayman Chemical Supplier No. 2402150 MG
A histamine H1 receptor antagonist (Ki = 10 nM in COS-7 cells expressing the human receptor); completely inhibits response to histamine in guinea pigs when administered at a dose of 1 mg/kg