Learn More
Accurate Chemical Acryl-AquaClean, Microbiological Additive for heating bath fluids (a WAK-Chemie product), 250ml
SDP
Catalog No. nc9918427
Encompass
Microbiological additive for heating bath fluids compatible with acrylic glass and aluminium. Inhibits growth of bacteria, algae and fungi. Beginning of contamination is indicated by decolorisation of the blue liquid. Active ingredients are non-volatile. 250ml
Catalog No. NC9918427 Supplier Accurate Chemical Supplier No. WAKAQA25050L
Microbiological additive for heating bath fluids compatible with acrylic glass and aluminium. Inhibits growth of bacteria, algae and fungi. Beginning of contamination is indicated by decolorisation of the blue liquid. Active ingredients are non-volatile. 250ml