Learn More
Research Products International Corp Acrylamide/bis-Acryl 19 1 500G
SDP
Catalog No. 502230274
Encompass_Preferred
Pre-measured powder blend of acrylamide and bis-acrylamide for simple stock solution preparation.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-223-0274 Supplier Research Products International Corp Supplier No. A11300500.0
Pre-measured powder blend of acrylamide and bis-acrylamide for simple stock solution preparation.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.