Learn More
Research Products International Corp Acrylamide/bis-Acryl Sol 5L
SDP
Catalog No. 502230278
Encompass_Preferred
A 40% solution containing 38.96% (w/v) Acrylamide and 1.04% (w/v) bis-Acrylamide for a monomer to crosslinker ratio of 37.5:1.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.
Catalog No. 50-223-0278 Supplier Research Products International Corp Supplier No. A114105000.0
A 40% solution containing 38.96% (w/v) Acrylamide and 1.04% (w/v) bis-Acrylamide for a monomer to crosslinker ratio of 37.5:1.
WARNING! This product can expose you to Acrylamide, a chemical which is known to the State of California to cause cancer, developmental and reproductive harm. For more information go to www.P65Warnings.ca.gov.