Learn More
National Diagnostics ACRYLAGEL 1 LT
SDP
Catalog No. 5089990063
Encompass
AcrylaGel, Ready-to-use, 30% acrylamide solution in distilled/deionized water. 18 megOhm water. 0.2 micron filtration, 1 LT
Catalog No. 50-899-90063 Supplier National Diagnostics Supplier No. EC8101LTR
AcrylaGel, Ready-to-use, 30% acrylamide solution in distilled/deionized water. 18 megOhm water. 0.2 micron filtration, 1 LT