Learn More
Alkali Scientific Acrylamide, 100 G
SDP
Supplier:  Alkali Scientific ACR001CP
Encompass_Preferred
Acrylamide is a chemical commonly used in electrophoresis, polymerization, and protein separation processes. The 100 g powder form is ideal for preparing gels for electrophoresis, particularly in research and quality control laboratories. Acrylamide polymerizes in the presence of a cross-linking agent to form polyacrylamide gels, which are used for separating proteins based on their size and charge. In addition to electrophoresis, acrylamide is used in the synthesis of other polymers and in laboratory techniques for DNA sequencing, protein analysis, and gel electrophoresis. This versatile reagent plays a key role in molecular biology and biochemistry applications, ensuring precise and reliable results in experimental procedures.
Catalog No. 50-006-60632