Learn More
Crescent Chemical Co Inc ACRYLAMIDE 2X RESEARCH GRADE
SDP
Catalog No. 50247126
Encompass_Preferred
Acrylamide 2X research grade Standard quality, applicable to general electrophoretic separations. Size - 1KG Storage Conditions - +2 °C TO +8 °C Catalog Number - 10675.02 CAS 79-06-1
Catalog No. 50-247-126 Supplier Crescent Chemical Co Inc Supplier No. SE10675.02
Acrylamide 2X research grade Standard quality, applicable to general electrophoretic separations. Size - 1KG Storage Conditions - +2 °C TO +8 °C Catalog Number - 10675.02 CAS 79-06-1